missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lipocalin-2/NGAL Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00 EUR - 2100.00 EUR
Specifications
| Antigen | Lipocalin-2/NGAL |
|---|---|
| Clone | 228421 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746468
![]() |
R&D Systems
MAB18571-SP |
25 μg |
209.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18714227
![]() |
R&D Systems
MAB18571-100 |
100 μg |
517.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18741698
![]() |
R&D Systems
MAB18571-500 |
500 μg |
2100.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| Lipocalin-2/NGAL | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
| LCN2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 228421 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P11672 | |
| 3934 | |
| NS0-derived recombinant mouse Lipocalin-2/NGAL Gln21-Asn200 Accession # P11672 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Lipocalin-2/NGAL in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) Lipocalin-1, rhLipocalin-2 or recombinant rat Lipocalin-2 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title