missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lipocalin-2/NGAL Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB18571-SP
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P11672 | |
| LCN2 | |
| NS0-derived recombinant mouse Lipocalin-2/NGAL Gln21-Asn200 Accession # P11672 | |
| 25 μg | |
| Primary | |
| Detects mouse Lipocalin-2/NGAL in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant human (rh) Lipocalin-1, rhLipocalin-2 or recombinant rat Lipocalin-2 is observed. | |
| Mouse | |
| Purified |
| ELISA | |
| 228421 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3934 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction