missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DcTRAILR2/TNFRSF22 Antibody, R&D Systems™

Rat Monoclonal Antibody
188.00 EUR - 385.00 EUR
Specifications
| Antigen | DcTRAILR2/TNFRSF22 |
|---|---|
| Clone | 263310 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755509
![]() |
R&D Systems
MAB1816-SP |
25 μg |
188.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18110411
![]() |
R&D Systems
MAB1816 |
100 μg |
385.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DcTRAILR2/TNFRSF22 Monoclonal specifically detects DcTRAILR2/TNFRSF22 in Mouse samples. It is validated for Western Blot.Specifications
| DcTRAILR2/TNFRSF22 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9ER62 | |
| Tnfrsf22 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 263310 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 8718 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R2/TNFRSF22 Ala39-Arg175 Accession # Q9ER62 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse DcTRAIL R2/TNFRSF22 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) 4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmDcTRAIL R1, rmDR3, rmEDAR, rmFas, rmGITR, rmHVEM, rmLT beta R, rmNGF R, rmOPG, rmOX40, rmRANK, rmTNF RI, and rmTNF RII is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title