missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DcTRAILR2/TNFRSF22 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1816-SP
This item is not returnable.
View return policy
Description
DcTRAILR2/TNFRSF22 Monoclonal specifically detects DcTRAILR2/TNFRSF22 in Mouse samples. It is validated for Western Blot.
Specifications
| DcTRAILR2/TNFRSF22 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9ER62 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8718 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 263310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Tnfrsf22 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R2/TNFRSF22 Ala39-Arg175 Accession # Q9ER62 | |
| 25 μg | |
| Primary | |
| Detects mouse DcTRAIL R2/TNFRSF22 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) 4-1BB, rmBAFF R, rmCD27, rmCD30, rmCD40, rmDcTRAIL R1, rmDR3, rmEDAR, rmFas, rmGITR, rmHVEM, rmLT beta R, rmNGF R, rmOPG, rmOX40, rmRANK, rmTNF RI, and rmTNF RII is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction