missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™

Rat Monoclonal Antibody
221.00 EUR - 464.00 EUR
Specifications
| Antigen | Angiopoietin-like Protein 4/ANGPTL4 |
|---|---|
| Clone | 547112 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773977
![]() |
R&D Systems
MAB9797-SP |
25 μg |
221.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744167
![]() |
R&D Systems
MAB9797-100 |
100 μg |
464.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Angiopoietin-like Protein 4/ANGPTL4 Monoclonal antibody specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Mouse samples. It is validated for ELISASpecifications
| Angiopoietin-like Protein 4/ANGPTL4 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein, Gln24-Ser410, Accession # Q9Z1P8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 547112 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9Z1P8 | |
| 51129 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title