Learn More
Fisherbrand™ Motorised Pipette Controller
Product Code. 15840053 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Motorised Pipette Controller
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15840053 Motorised Pipette Controller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15840053
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Edge
Add to basket
A powerful, lightweight and ergonomically designed pipetting controller for all commonly used glass or plastic pipettes.
  • Powerful yet quiet pump
  • Ergonomic design
  • Soft grip push buttons
  • Double safety valve
  • Autoclavable pipette cone
  • two speed modes High, Low and Gravity
  • Switch located on slide part of controller ensuring ease of operation
  • LED indicator lights up in red when battery is low
  • Can be operated continuously for 8 hours
  • Replaceable membrane Filter to protect device from contaminants being drawn into pipette
  • Can be used while it is charging
Pipet, filler, liquid handling
Specifications
Battery Type 1000mAh NiMH batteries
Compatible Tips Cone shape
Color Blue
Grip Type Soft Push Button
Type Pipette Controller
Volume (Metric) 0.1 to 200 mL
Filter Type Hydrophobic PTFE membrane filter
Finger Style Push button
For Use With (Equipment) Glass or plastic serological pipettes
Includes Pipette filler, Charger (EU plug), Desktop stand, Wall mount bracket, Two spare membrane filters: 0.20μm and 0.45μm, Instruction manual, Batteries
Number of Speeds Two (High and Low, along with Gravity drain)
Recharge Time 2 to 3 hours
Electrical Requirements 2 x 1.2V / 1000mAh NiMH batteries
Voltage 1.2 V