missing translation for 'onlineSavingsMsg'
Learn More

Motolimod, MedChemExpress

Product Code. 16495848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495848 10 mg 10mg
16458507 5 mg 5mg
16453386 25 mg 25mg
16491997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495848 Supplier Medchem Express Supplier No. HY13773

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Motolimod (VTX-2337;VTX-378) is a selective Toll-like receptor 8 (TLR8) agonist, with an EC50 of approximately 100 nM.

Motolimod (VTX-2337

Specifications

Chemical Name or Material Motolimod
Physical Form Solid
CAS 926927-61-9
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C28H34N4O2
For Use With (Application) Cancer-programmed cell death
Synonym VTX-2337 VTX-378
Solubility Information DMSO : 50 mg/mL (109.03 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC=C(C3=CC=C(C(N4CCCC4)=O)C=C3)C=C2N=C(N)C1)N(CCC)CCC
Molecular Weight (g/mol) 458.6
Formula Weight 458.6
Percent Purity 97.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.