Learn More
Mosapride citrate, MedChemExpress
Product Code. 16438398
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16438398
Brand: Medchem Express HYB0189A
MedChemExpress Mosapride citrate is a gastroprokinetic agent that acts as a selective 5HT4 agonist.
Mosapride citrate is a gastroprokinetic agent that acts as a selective 5HT4 agonist.
Specifications
Chemical Name or Material Mosapride citrate
CAS 112885-42-4
Color White
Purity Grade Notes Research
Molecular Formula C27H33ClFN3O10
Quantity 10 mg
Synonym TAK-370 citrate AS-4370 citrate
Solubility Information DMSO : 100 mg/mL (162.86 mM; Need ultrasonic)
SMILES O=C(NCC1CN(CC2=CC=C(F)C=C2)CCO1)C3=CC(Cl)=C(N)C=C3OCC.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 614.02
Formula Weight 614.02
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation