missing translation for 'onlineSavingsMsg'
Learn More

Mosapride Citrate Dihydrate, TRC

Product Code. 30136496
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136496 50 mg 50mg
30007348 1 g 1g
17951985 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136496 Supplier Toronto Research Chemicals Supplier No. TRCM73099550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636582-62-2
Molecular Formula C21 H25 Cl F N3 O3 . C6 H8 O7 . 2 H2 O
Molecular Weight (g/mol) 650.05
Synonym Benzamide, 4-amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-2-morpholinyl]methyl]-, 2-hydroxy-1,2,3-propanetricarboxylate, hydrate (1:1:2), Benzamide, 4-amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-2-morpholinyl]methyl]-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), dihydrate (9CI), Mosapride citrate dihydrate, 4-Amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]-2-morpholinyl]methyl]benzamide citrate dihydrate, 4-Amino-5-chloro-2-ethoxy-N-[[4-(4-fluorobenzyl)-2-morpholinyl]methyl]benzamide citrate dihydrate
IUPAC Name 4-amino-5-chloro-2-ethoxy-N-[[4-[(4-fluorophenyl)methyl]morpholin-2-yl]methyl]benzamide;2-hydroxypropane-1,2,3-tricarboxylic acid;dihydrate
SMILES O.O.CCOc1cc(N)c(Cl)cc1C(=O)NCC2CN(Cc3ccc(F)cc3)CCO2.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H25ClFN3O3.C6H8O7.2H2O/c1-2-28-20-10-19(24)18(22)9-17(20)21(27)25-11-16-13-26(7-8-29-16)12-14-3-5-15(23)6-4-14;7-3(8)1-6(13,5(11)12)2-4(9)10;;/h3-6,9-10,16H,2,7-8,11-13,24H2,1H3,(H,25,27);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H2
Formula Weight 649.205
Percent Purity >95%
Chemical Name or Material Mosapride Citrate Dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.