missing translation for 'onlineSavingsMsg'
Learn More

Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Thermo Scientific™

Product Code. p-3586850
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10238903 1 g Amber Glass Bottle 1g
10602685 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10238903 Supplier Thermo Scientific Maybridge Supplier No. MO08372DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 173341-02-1
Molecular Formula C10H20N2O3
Molecular Weight (g/mol) 216.28
MDL Number MFCD06808588,MFCD12545849,MFCD12545850
InChI Key IYHJNCQAADULQE-UHFFFAOYNA-N
Synonym tert-butyl morpholin-2-ylmethyl carbamate, tert-butyl morpholin-2-ylmethylcarbamate, 2-n-boc-aminomethylmorpholine, tert-butyl n-morpholin-2-ylmethyl carbamate, tert-butyl morpholin-2-yl methyl carbamate, morpholin-2-ylmethyl-carbamic acid tert-butyl ester, carbamic acid,n-2-morpholinylmethyl-, 1,1-dimethylethyl ester, carbamic acid, n-2-morpholinylmethyl-, 1,1-dimethylethyl ester, n-boc-morpholine-2-methanamine, 2-n-boc-aminomethyl morpholine
PubChem CID 19081107
SMILES CC(C)(C)OC(=O)NCC1CNCCO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
IUPAC Name tert-butyl N-(morpholin-2-ylmethyl)carbamate
Quantity 1 g
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.