Learn More
Moroxydine-d8 Hydrochloride, TRC
Product Code. 30134746
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134746 10 mg 10mg
17997659 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30134746 Supplier Toronto Research Chemicals Supplier No. TRCM65175210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469734-82-3
Molecular Formula C6H5D8N5O .HCl
Molecular Weight (g/mol) 215.71
Synonym N-[Imino(4-morpholinyl)methyl]guanidine Hydrochloride-d8, N1,N1-Anhydrobis(β-hydroxyethyl)biguanide Hydrochloride-d8, SKF 8898A Hydrochloride-d8, Vironil Hydrochloride-d8, Virugon Hydrochloride-d8, Virumin Hydrochloride-d8, Wirumin Hydrochloride-d8, 4-Morpholinecarboximidoylguanidine Hydrochloride-d8, ABOB Hydrochloride-d8, JD 1022 Hydrochloride-d8, N-Amidino-4-morpholinecarboxamidine Hydrochloride-d8, N-(Aminoiminomethyl)-4-morpholinecarboximidamide Hydrochloride-d8
IUPAC Name N-carbamimidoylmorpholine-d8-4-carboximidamide hydrochloride
SMILES N=C(NC(N)=N)N1C([2H])([2H])C([2H])([2H])OC([2H])([2H])C1([2H])[2H].Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C6H13N5O.ClH/c7-5(8)10-6(9)11-1-3-12-4-2-11;/h1-4H2,(H5,7,8,9,10);1H/i1D2,2D2,3D2,4D2;
Formula Weight 215.14
Percent Purity >95%
Chemical Name or Material Moroxydine-d8 Hydrochloride
Grade HPLC