Learn More
Morantel Citrate, TRC
Product Code. 30011570
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011570 1 g 1g
30009590 250 mg 250mg
17972455 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011570 Supplier Toronto Research Chemicals Supplier No. TRCM6303531G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69525-81-1
Molecular Formula C12H16N2S • C6H8O7
Molecular Weight (g/mol) 220.3319212
Synonym 1,4,5,6-Tetrahydro-1-methyl-2-[(1E)-2-(3-methyl-2-thienyl)ethenyl]pyrimidine 2-Hydroxy-1,2,3-propanetricarboxylate;
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;1-methyl-2-[(E)-2-(3-methylthiophen-2-yl)ethenyl]-5,6-dihydro-4H-pyrimidine
SMILES CC(C=CS1)=C1/C=C/C2=NCCCN2C.O=C(CC(C(O)=O)(O)CC(O)=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C12H16N2S.C6H8O7/c1-10-6-9-15-11(10)4-5-12-13-7-3-8-14(12)2;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-6,9H,3,7-8H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b5-4+;
Formula Weight 412.1304
Chemical Name or Material Morantel Citrate