missing translation for 'onlineSavingsMsg'
Learn More

MOPSO sodium salt

Product Code. 16584214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16584214 500 g 500g
16564214 25 g 25g
16574214 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16584214 Supplier Thermo Scientific Acros Supplier No. 462145000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79803-73-9 | C7H14NNaO5S | 247.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.24
InChI Key WSFQLUVWDKCYSW-UHFFFAOYNA-M
IUPAC Name sodium 2-hydroxy-3-(morpholin-4-yl)propane-1-sulfonate
SMILES [Na+].OC(CN1CCOCC1)CS([O-])(=O)=O

Specifications

Quantity 500 g
Solubility Information Solubility aq. soln.: 500 g/L
Formula Weight 247.24
Chemical Name or Material MOPSO sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.