Learn More
Montelukast Bis-sulfide (mixture of diastereomers), TRC
Product Code. 30136180
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136180 100 mg 100mg
30008885 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136180 Supplier Toronto Research Chemicals Supplier No. TRCM567985100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1242260-05-4
Molecular Formula C41 H46 Cl N O5 S2
Molecular Weight (g/mol) 732.39
Synonym 1-[[[(1R)-1-[3-[(1RS)-1-[[[1-(Carboxymethyl)cyclopropyl]methyl]sulfanyl]-2-(7-chloroquinolin-2-yl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid, Montekulast Sodium Imp. D+E (Mixture of Stereoisomers) (EP), 1-[[[(1R)-1-[3-[1-[[[1-(Carboxymethyl)cyclopropyl]methyl]thio]-2-(7-chloro-2-quinolinyl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic Acid
IUPAC Name 2-[1-[[(1R)-1-[3-[1-[[1-(carboxymethyl)cyclopropyl]methylsulfanyl]-2-(7-chloroquinolin-2-yl)ethyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid
SMILES CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(c3)C(Cc4ccc5ccc(Cl)cc5n4)SCC6(CC(=O)O)CC6
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C41H46ClNO5S2/c1-39(2,48)33-9-4-3-6-27(33)12-15-35(49-25-40(16-17-40)23-37(44)45)29-7-5-8-30(20-29)36(50-26-41(18-19-41)24-38(46)47)22-32-14-11-28-10-13-31(42)21-34(28)43-32/h3-11,13-14,20-21,35-36,48H,12,15-19,22-26H2,1-2H3,(H,44,45)(H,46,47)/t35-,36?/m1/s1
Formula Weight 731.2506
Chemical Name or Material Montelukast Bis-sulfide (mixture of diastereomers)