Learn More
TFA Dostmann™ Monsun Wireless Rain Gauge
Product Code. 11897870
Change view
Click to view available options
Temperature (Metric) Operating:
-50°C to +70°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Operating Unit Size
11897870 -50°C to +70°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11897870 Supplier TFA Dostmann™ Supplier No. 47.3003
  • ONLINE EXCLUSIVE PRICE
    43.77 EUR
    32.13 EUR / Pack of 1
    Save 11.64 EUR
    26% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Specifications
For Use With (Application) Wireless transmission of outdoor temperature and rainfall amounts (max. 30 m) with temperature trend
Height (Metric) 122 mm
Material Plastic
Temperature (Metric) Operating -50°C to +70°C
Weight (Metric) 114 g
Product Type Wireless Rain Gauge