Learn More
Monopentyl Phthalate, TRC
Product Code. 30136899
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136899 250 mg 250mg
30006132 50 mg 50mg
30007449 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136899 Supplier Toronto Research Chemicals Supplier No. TRCM566740250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24539-56-8
Molecular Formula C13 H16 O4
Molecular Weight (g/mol) 236.26
Synonym Phthalic acid mono-n-pentyl ester
IUPAC Name 2-pentoxycarbonylbenzoic acid
SMILES CCCCCOC(=O)c1ccccc1C(=O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C13H16O4/c1-2-3-6-9-17-13(16)11-8-5-4-7-10(11)12(14)15/h4-5,7-8H,2-3,6,9H2,1H3,(H,14,15)
Formula Weight 236.105
Percent Purity >95%
Chemical Name or Material Mono-n-pentyl phthalate
Grade HPLC