Learn More
Monohexyl Phthalate, TRC
Product Code. 30009799
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009799 10 g 10g
30011620 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009799 Supplier Toronto Research Chemicals Supplier No. TRCM54648010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24539-57-9
Molecular Formula C14 H18 O4
Molecular Weight (g/mol) 250.29
Synonym Phthalic acid, mono-hexyl ester
IUPAC Name 2-hexoxycarbonylbenzoic acid
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C14H18O4/c1-2-3-4-7-10-18-14(17)12-9-6-5-8-11(12)13(15)16/h5-6,8-9H,2-4,7,10H2,1H3,(H,15,16)
Formula Weight 250.1205
Percent Purity >95%
Chemical Name or Material Phthalic acid, mono-hexyl ester
Grade HPLC