Learn More
Monohexyl Phthalate-d4, TRC
Product Code. 30135046
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135046 50 mg 50mg
17938509 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30135046 Supplier Toronto Research Chemicals Supplier No. TRCM54648250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469279-12-5
Molecular Formula C14H14D4O4
Molecular Weight (g/mol) 254.31
Synonym 1,2-Benzenedicarboxylic Acid Monohexyl Ester-d4, Phthalic Acid Monohexyl Ester-d4, Flexol DHD-d4, Mono-n-hexyl Phthalate-d4, Monohexyl Phthalate-d4, n-Hexyl Acid Phthalate-d4
SMILES O=C(OCCCCCC)C1=C([2H])C([2H])=C([2H])C([2H])=C1C(O)=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H18O4/c1-2-3-4-7-10-18-14(17)12-9-6-5-8-11(12)13(15)16/h5-6,8-9H,2-4,7,10H2,1H3,(H,15,16)/i5D,6D,8D,9D
Percent Purity >95%
Chemical Name or Material Monohexyl Phthalate-d4
Grade HPLC