Learn More
Monoheptyl Phthalate, TRC
Specifications
Specifications
| Chemical Name or Material | Monoheptyl Phthalate |
| CAS | 24539-58-0 |
| Molecular Formula | C15H20O4 |
| Quantity | 100 mg |
| Synonym | 1-Heptyl Ester 1,2-Benzenedicarboxylic Acid, Phthalic Acid Heptyl Ester, Phthalic Acid Monoheptyl Ester |
| InChI Formula | InChI=1S/C15H20O4/c1-2-3-4-5-8-11-19-15(18)13-10-7-6-9-12(13)14(16)17/h6-7,9-10H,2-5,8,11H2,1H3,(H,16,17) |
| SMILES | CCCCCCCOC(=O)c1ccccc1C(=O)O |
| IUPAC Name | 2-heptoxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 264.32 |
| Formula Weight | 264.1362 |