Learn More
Mono-O-acetyl Fingolimod TFA, TRC
Specifications
Specifications
| Chemical Name or Material | Mono-O-acetyl Fingolimod TFA |
| CAS | 2469039-45-8 |
| Molecular Formula | C21H35NO3 . C2HF3O2 |
| Quantity | 100 mg |
| Synonym | 2-Amino-2-[2-(4-octylphenyl)ethyl]-1,3-propanediol 1-Acetate TFA |
| InChI Formula | InChI=1S/C21H35NO3.C2HF3O2/c1-3-4-5-6-7-8-9-19-10-12-20(13-11-19)14-15-21(22,16-23)17-25-18(2)24;3-2(4,5)1(6)7/h10-13,23H,3-9,14-17,22H2,1-2H3;(H,6,7) |
| SMILES | NC(COC(C)=O)(CO)CCC1=CC=C(CCCCCCCC)C=C1.OC(C(F)(F)F)=O |
| IUPAC Name | 2-amino-2-(hydroxymethyl)-4-(4-octylphenyl)butyl acetate 2,2,2-trifluoroacetate |
| Molecular Weight (g/mol) | 463.53 |
| Formula Weight | 463.25 |