Learn More
Mono-2-ethylHydroxyhexyl Terephthalate (MEHHTP), TRC
Product Code. 17922205
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922205 10 mg 10mg
17931865 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17922205 Supplier Toronto Research Chemicals Supplier No. TRCM52565010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1684398-38-6
Molecular Formula C16H22O5
Molecular Weight (g/mol) 294.34
Synonym 1,4-Benzenedicarboxylic Acid 1-(2-Ethyl-5-hydroxyhexyl) Ester, MEHHTP;
IUPAC Name 4-(2-ethyl-5-hydroxy-hexoxy)carbonylbenzoic acid
SMILES CCC(CCC(C)O)COC(=O)c1ccc(cc1)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H22O5/c1-3-12(5-4-11(2)17)10-21-16(20)14-8-6-13(7-9-14)15(18)19/h6-9,11-12,17H,3-5,10H2,1-2H3,(H,18,19)
Formula Weight 294.1467
Percent Purity >95%
Chemical Name or Material Mono-2-ethylHydroxyhexyl Terephthalate
Grade HPLC