Learn More
Mono(2-ethylhexyl) Terephthalate, TRC
Product Code. 30135353
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135353 250 mg 250mg
30136674 5 g 5g
30011628 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135353 Supplier Toronto Research Chemicals Supplier No. TRCM542570250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 155603-50-2
Molecular Formula C16 H22 O4
Molecular Weight (g/mol) 278.34
Synonym 1,4-Benzenedicarboxylic acid, mono(2-ethylhexyl) ester (9CI), 2-Ethylhexyl terephthalate, Mono(2-ethylhexyl) terephthalate, Mono(2-Ethylhexyl) Terephthalate, Terephthalic acid, mono-2-ethylhexyl ester
IUPAC Name 4-(2-ethylhexoxycarbonyl)benzoic acid
SMILES CCCCC(CC)COC(=O)c1ccc(cc1)C(=O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C16H22O4/c1-3-5-6-12(4-2)11-20-16(19)14-9-7-13(8-10-14)15(17)18/h7-10,12H,3-6,11H2,1-2H3,(H,17,18)
Formula Weight 278.1518
Percent Purity >95%
Chemical Name or Material Mono-2-ethylhexyl-terephthalate
Grade HPLC