Learn More
MilliporeSigma Molybdenum(VI) Oxide, For Analysis, EMSURE™, MilliporeSigma™
Product Code. 15863880
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15863880 500 g Plastic bottle 500g
15853880 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15863880 Supplier MilliporeSigma Supplier No. 1004030500
 Restricted Item
Estimated Shipment: 19-11-2026
CAS: 1313-27-5 | MoO3 | 143.95 g/mol
For analysis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O
Specifications
Melting Point 795°C
Color Yellow
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.5%
Linear Formula MoO3
Solubility Information Solubility in Water - 0.49 g/L
InChI Formula InChI=1S/Mo.3O
Formula Weight 143.94
Percent Purity ≥99.5%
Concentration or Composition (by Analyte or Components) Molybdenum(VI) oxide (MoO3) ≥99.5%
Manufacturing Origin Germany
Odor Odorless
Packaging Plastic bottle
Product Line EMSURE
Appearance Yellow solid
Density 4.70 g/cm³
Assay Precipitation Titration
Chemical Name or Material Molybdenum(VI) oxide