Learn More
Molidustat Sodium Salt, TRC
Product Code. 30012698
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012698 25 mg 25mg
30009207 1 mg 1mg
30007905 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012698 Supplier Toronto Research Chemicals Supplier No. TRCM48745025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1375799-59-9
Molecular Formula C13H13N8NaO2
Molecular Weight (g/mol) 336.28
Synonym 1-[6-(4-Morpholinyl)-4-pyrimidinyl]-4-(1H-1,2,3-triazol-1-yl)-1H-pyrazol-5-ol Sodium Salt
SMILES [Na]OC1C(N2C=CN=N2)=CNN1C3=CC(N4CCOCC4)=NC=N3
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C13H14N8O2.Na/c22-13-10(20-2-1-16-18-20)8-17-21(13)12-7-11(14-9-15-12)19-3-5-23-6-4-19;/h1-2,7-9,22H,3-6H2;/q;+1/p-1
Formula Weight 336.1059
Percent Purity >95%
Chemical Name or Material Molidustat Sodium Salt
Grade HPLC