Learn More
Invitrogen™ Thiol and Sulfide Quantitation Kit
Product Code. 10388252
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10388252 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10388252 Supplier Invitrogen™ Supplier No. T6060
Ultrasensitive colorimetric assay for quantifying both protein and nonprotein thiols
In this assay, thiols or inorganic sulfides reduce a disulfide-inhibited derivative of papain, stoichiometrically releasing the active enzyme. The activity of the enzyme is then measured using the chromogenic papain substrate N-benzoyl-L-arginine, p-nitroanilide (L-BAPNA). The enzymatic amplification step in this kit enables researchers to detect as little as 0.2μM thiol - a sensitivity that is about 100-fold greater than that achieved using Ellman's reagent.
Anion Detection, Biochemical Toxicology Assays, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Product Type Thiol and Sulfide Quantitation Kit
Quantity 1 Kit
compliance-icons
Product Identifier
  • Thiol and Sulfide Quantitation Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cystamine dihydrochloride