Learn More
Invitrogen™ Molecular Probes™ ProLong™ Antifade Kit
Product Code. 10720764
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10720764 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10720764 Supplier Invitrogen™ Supplier No. P7481
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Allows longer-term storage of the sample
The ProLong Antifade Kit provides a powdered antifade reagent and a liquid mounting medium that can be mixed together and applied directly to fluorescently labeled cell or tissue samples on microscope slides to protect the dyes from fading (photobleaching) during fluorescence microscopy experiments.
  • Protects dyes from fading during imaging
  • Mix powdered antifade with mounting medium and use the same day
  • Mounted samples are stable for months
  • Offers excellent photobleaching protection for Texas Red™ and most Alexa Fluor™ dyes
  • Liquid mounting medium component can be used without antifade reagent for acceptor photobleaching FRET experiments
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Quantity 1 kit