Learn More
Invitrogen™ Marina Blue™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine (Marina Blue™ DHPE)
Product Code. 10207592
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10207592 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10207592 Supplier Invitrogen™ Supplier No. M12652
 Restricted Item
In Stock
A phospholipid
Marina Blue DHPE is labeled on the head group with the blue-fluorescent Marina Blue fluorophore with excitation/emission maxima ∽365/460nm.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phospholipids
Quantity 1 mg
Physical Form Solid
Product Line MARINA BLUE