missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Molecular Probes™ H2DIDS (H2DIDS
Description
H2DIDS can be used to inhipit anionic transport.
Cell-Based Ion Channel Assays, Drug Discovery & Development, Ion Channel Biology, Target & Lead Identification & Validation
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 100 mg |
| Color | Ultraviolet |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?