missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiBAC4(3) (Bis-(1,3-Dibutylbarbituric Acid)Trimethine Oxonol)

Product Code. 11504197
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504197 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504197 Supplier Invitrogen™ Supplier No. B438
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The slow-response potential-sensitive probe, DiBAC4(3) can enter depolarized cells where it binds to intracellular proteins or membrane and exhibits enhanced fluorescence and a red spectral shift.

The slow-response potential-sensitive probe, DiBAC4(3) can enter depolarized cells where it binds to intracellular proteins or membrane and exhibits enhanced fluorescence and a red spectral shift. Increased depolarization results in additional influx of the anionic dye and an increase in fluorescence. Conversely, hyperpolarization is indicated by a decrease in fluorescence. This bis-oxonal has an excitation maxima of 490 nm and emission maxima of 516 nm. DiBAC dyes are excluded from mitochondria because of their overall negative charge, making them superior to carbocyanines for measuring plasma membrane potentials.

Dissolve in high-quality anhydrous DMSO or ethanol to prepare a stock concentration up to 1 mM.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature and protect from light.
Product Type DiBAC4(3)
Dye Type Membrane Potential Probes
Quantity 25 mg
Shipping Condition Room Temperature
Excitation/Emission 493/516 nm
Molecular Weight (g/mol) 516.64

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.