Learn More
Invitrogen™ CholEsteryl 4,4-Difluoro-5-(4-Methoxyphenyl)-4-Bora-3a,4a-Diaza-s-Indacene-3-Undecanoate (CholEsteryl BODIPY™ 542/563 C11)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Cholesteryl esters consist of a fatty acid esterified to the 3β-hydroxyl group of cholesterol. These very nonpolar species are the predominant lipid components of atherosclerosis plaque and low- and high-density lipoporotein (LDL and HDL) cores.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Cholesterol Analogs |
| Quantity | 1 mg |
| Physical Form | Solid |
| Product Line | BODIPY |