Learn More
Invitrogen™ BODIPY™ FL DHPE (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Product Code. 11520316
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11520316 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11520316 Supplier Invitrogen™ Supplier No. D3800
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Can form excimers at high concentrations
BODIPY DHPE has the green-fluorescent dye attached to the head group of the phospholipid. Unlike fluorophores like pyrene, DPH and NBD, BODIPY FL is relatively environment insensitive and is fluorescent in both aqueous and lipid environments.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phospholipids
Quantity 100 μg
Physical Form Solid
Product Line BODIPY