Learn More
Invitrogen™ BODIPY™ FL C5-Ceramide complexed to BSA
Product Code. 11584167
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11584167 5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11584167 Supplier Invitrogen™ Supplier No. B22650
  • ONLINE EXCLUSIVE PRICE
    678.00 EUR
    616.65 EUR / 5mg
    Save 61.35 EUR
    9% Off
View all promotions
 Restricted Item
In Stock
Can be used in sphingolipid transport and metabolism mechanisms
Complexing fluorescent lipids with bovine serum albumin (BSA) facilitates cell labeling by eliminating the need for organic solvents to dissolve the lipophilic probe - the BSA-complexed probe can be directly dissolved in water.
Cell Analysis, Cell Structure, Golgi
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sphingolipids
Quantity 5 mg
Physical Form Solid
Product Line BODIPY