Learn More
Invitrogen™ BODIPY™ FL C5 (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoic Acid)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
The green fluorescent fatty acid, BODIPY FL C5 can be used as a synthetic precursor to a wide variety of fluorescent phospholipids. Unlike fluorophores like pyrene, DPH and NBD, BODIPY dyes are relatively environment insensitive.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Fatty Acids |
| Quantity | 1 mg |
| Physical Form | Solid |
| Product Line | BODIPY |