Learn More
Invitrogen™ BODIPY™ 500/510 C1, C12 (4,4-Difluoro-5-Methyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoic Acid)
Product Code. 10297622
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 10297622
Brand: Invitrogen™ D3823
Edge
Forms excimers and exhibits red-shifted fluorescence emission when incorporated into living cells
C1-BODIPY 500/510 C12 is a fluorescent fatty acid analog that has been used for lipid trafficking studies and as a general membrane probe.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fatty Acids
Quantity 1 mg
Physical Form Solid
Product Line BODIPY