Learn More
Invitrogen™ β-BODIPY™ FL C12-HPC (2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Dodecanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphocholine)
Description
The intensely fluorescent BODIPY (4,4-difluoro-3a,4a-diaza-
s-indacene) fluorophore is intrinsically lipophilic, unlike most other long-wavelength dyes. Consequently, probes incorporating this fluorophore are more likely to mimic the properties of natural lipids. BODIPY™ fatty acid analogs like BODIPY 500/510 dodecanoic acid (D-3823) and BODIPY FL dodecanoic acid (D-3822) undergo native-like transport and metabolism in cells.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Phospholipids |
| Quantity | 100 μg |
| Physical Form | Solid |
| Product Line | BODIPY |