Learn More
Invitrogen™ Amplex™ Red Uric Acid/Uricase Assay Kit
Description
Detects uric acid or uricase using either a fluorescence microplate reader or fluorometer
The Amplex Red Uric Acid/Uricase Assay Kit provides a sensitive and simple method for detecting uric acid or uricase using either a fluorescence microplate reader or fluorometer. Serum uric acid is the end product of purine metabolism in the body tissues and is cleared through the kidneys by glomerular filtration. Most animals can metabolize uric acid to more readily excreted products, but humans lack the necessary enzyme, urate oxidase (uricase), as a result of the presence of two 'nonsense mutations' in the human gene for uricase. The Amplex Red Uric Acid/Uricase Assay Kit provides an ultrasensitive method for detecting uric acid or for monitoring uricase activity. In the assay, uricase catalyzes the conversion of uric acid to allantoin, hydrogen peroxide, and carbon dioxide. In the presence of horseradish peroxidase (HRP), hydrogen peroxide reacts stoichiometrically with the Amplex Red reagent to generate the red-fluorescent oxidation product resorufin. Because resorufin has absorption and fluorescence emission maxima of approximately 571nm and 585nm, respectively, there is little interference from autofluorescence in most biological samples.
- Detect concentrations as low as 100nm uric acid or 0.2mU/mL of uricase activity
- Format allows for multiple time point measurements
- Designed for minimal autofluorescence interference
- Custom assay design and packaging are also available
Use Amplex Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex UltraRed Reagent, a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex Red/UltraRed Stop Reagent. The Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours.
Enzyme and Protein Activity Assays, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Uric Acid/Uricase Assay |
| For Use With (Equipment) | Microplate Reader, Spectrophotometer, Fluorometer |
| Product Type | Amplex Red Assay Kit |
| Dye Type | Other Label(s) or Dye(s) |
| Format | Tube(s), 96-well plate |
| Product Line | Amplex |
| Quantity | 400 Assays |
| Shipping Condition | Room Temperature |