Learn More
Moexipril Diketopiperazine, TRC
Product Code. 30479232
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479232 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30479232 Supplier Toronto Research Chemicals Supplier No. TRCM485340100MG
 Restricted Item
Estimated Shipment: 09-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-ethyl 2-{(3S,11aS)-8,9-dimethoxy-3-methyl-1,4-dioxo-3,4-dihydro-1H-pyrazino[1,2-b]isoquinolin-2(6H,11H,11aH)-yl}-4-phenylbutanoate
CAS 103733-51-3
Molecular Formula C27 H32 N2 O6
Quantity 100 mg
Synonym (αS,3S,11aS)-1,3,4,6,11,11a-Hexahydro-8,9-dimethoxy-3-methyl-1,4-dioxo-α-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetic Acid Ethyl Ester, [3S-[2(R*),3α,11aβ]]-1,3,4,6,11,11a-Hexahydro-8,9-dimethoxy-3-methyl-1,4-dioxo-α-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetic Acid Ethyl Ester, PD 114009, Moexipril USP Related Compound B
InChI Formula InChI=1S/C27H32N2O6/c1-5-35-27(32)21(12-11-18-9-7-6-8-10-18)29-17(2)25(30)28-16-20-15-24(34-4)23(33-3)14-19(20)13-22(28)26(29)31/h6-10,14-15,17,21-22H,5,11-13,16H2,1-4H3
SMILES CCOC(=O)C(CCc1ccccc1)N2C(C)C(=O)N3Cc4cc(OC)c(OC)cc4CC3C2=O
IUPAC Name ethyl 2-(8,9-dimethoxy-3-methyl-1,4-dioxo-3,6,11,11a-tetrahydropyrazino[1,2-b]isoquinolin-2-yl)-4-phenylbutanoate
Molecular Weight (g/mol) 480.55
Formula Weight 480.226