Learn More
Moclobemide, MedChemExpress
Product Code. 16448356
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448356 100 mg 100mg
16444387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16448356 Supplier Medchem Express Supplier No. HYB0534
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A.Moclobemide up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice.
Moclobemide (Ro111163) is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A.Moclobemide up-regulates proliferation of hippocampal progenitor cells in chronically stressed mice.
Specifications
Chemical Name or Material Moclobemide
Physical Form Solid
CAS 71320-77-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H17ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym Ro111163
Solubility Information DMSO : 100 mg/mL (372.11 mM; Need ultrasonic)
SMILES O=C(NCCN1CCOCC1)C2=CC=C(Cl)C=C2
Molecular Weight (g/mol) 268.74
Formula Weight 268.74
Percent Purity 99.63%
Grade Research