missing translation for 'onlineSavingsMsg'
Learn More

MOBS, 0.2M buffer soln., pH 8.0, Thermo Scientific™

Product Code. 15465789
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 15465789

Brand: Thermo Scientific Alfa Aesar J63594.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115724-21-5
Molecular Formula C8H17NO4S
Molecular Weight (g/mol) 223.29
MDL Number MFCD00674916
InChI Key VTOWJTPBPWTSMK-UHFFFAOYSA-N
Synonym 4-(4-Morpholinyl)butanesulfonic acid
PubChem CID 3674064
IUPAC Name 4-morpholin-4-ylbutane-1-sulfonic acid
SMILES OS(=O)(=O)CCCCN1CCOCC1

Specifications

Color Colorless
Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material MOBS buffer soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.