accessibility menu, dialog, popup

UserName

CAS RN 115724-21-5

ONSOOHO

CAS RN 115724-21-5

IUPAC Name: 4-(morpholin-4-yl)butane-1-sulfonic acid
Synonyms: 4-(morpholin-4-yl)butane-1-sulfonic acid
Molecular Weight (g/mol): 223.29
Molecular Formula: C8H17NO4S
InChi Key: VTOWJTPBPWTSMK-UHFFFAOYSA-N
SMILES: OS(=O)(=O)CCCCN1CCOCC1
Quantity 
  • (4)
  • (4)
pH 
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (8)
  • (8)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)