Learn More
EKASTU Safety Mobile Wall-Cabinet for Masks
Product Code. 10131101
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10131101 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10131101 Supplier EKASTU Safety Supplier No. 266921
Estimated Shipment: 27-11-2026
EKASTU Mobile Wall-Cabinet for Masks is dust-proof for hygienic storage
  • Wall-mounting
  • Can be removed for transport
  • Shock and impact-resistant
  • Transparent door and 2 inserted shelves
  • Holds 1 full face mask or 3 half masks, each with filter
Specifications
Product Type Mobile wall-cabinet for masks
Color Green
Quantity 1 Ea.
Dimensions (L x W x H) 33 x 15 x 42 cm
Weight (Metric) Approx. 1.6 kg