Learn More
- Home
- Products
- Furniture
- Desks and Tables
- Laboratory Benches and Tables
- Köttermann Mobile Single Bench
Köttermann Mobile Single Bench
Product Code. 17392229
Change view
Click to view available options
Dimensions (D x W x H):
600 x 900 x 750 mm
600 x 900 x 900 mm
750 x 900 x 750 mm
750 x 900 x 900 mm
600 x 1200 x 750 mm
600 x 1200 x 900 mm
600 x 1500 x 750 mm
600 x 1500 x 900 mm
750 x 1200 x 750 mm
750 x 1200 x 900 mm
750 x 1500 x 750 mm
750 x 1500 x 900 mm
Top Style:
Melamine TopResist Worktop
Trespa TopLab PLUS Worktop
Unit Size:
Each
22 product options available for selection
| Product Code. | Dimensions (D x W x H) | Top Style | unitSize |
|---|---|---|---|
| 17392229 | 750 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17392199 | 600 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17322229 | 600 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17322209 | 600 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17352229 | 600 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17302209 | 600 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17332229 | 600 x 1200 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17362229 | 600 x 1200 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17312209 | 600 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17342229 | 600 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17332209 | 600 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17372229 | 600 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17342209 | 750 x 900 x 750 mm | Melamine TopResist Worktop | Each |
| 17382229 | 750 x 900 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17372209 | 750 x 900 x 900 mm | Melamine TopResist Worktop | Each |
| 17312239 | 750 x 900 x 900 mm | Trespa TopLab PLUS Worktop | Each |
| 17352209 | 750 x 1200 x 750 mm | Melamine TopResist Worktop | Each |
| 17382209 | 750 x 1200 x 900 mm | Melamine TopResist Worktop | Each |
| 17362209 | 750 x 1500 x 750 mm | Melamine TopResist Worktop | Each |
| 17302239 | 750 x 1500 x 750 mm | Trespa TopLab PLUS Worktop | Each |
| 17392209 | 750 x 1500 x 900 mm | Melamine TopResist Worktop | Each |
| 17322239 | 750 x 1500 x 900 mm | Trespa TopLab PLUS Worktop | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Available for sitting and standing work height
- Features castors for mobility
Specifications
Specifications
| Dimensions (D x W x H) | 750 x 1200 x 750 mm |
| No. of Shelves | 1 |
| Top Style | Trespa TopLab PLUS Worktop |