Learn More
Miele Professional™ Mobile Injector
Product Code. 13172290 Shop All Miele Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13172290 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13172290 Supplier Miele Professional™ Supplier No. 3809340
  • ONLINE EXCLUSIVE PRICE
    2365.00 EUR
    1521.62 EUR / Pack of 1
    Save 843.38 EUR
    35% Off
Estimated Shipment: 23-12-2026
Mobile unit for the optimum loading of narrow necked glassware
  • Equipped with 39 injector nozzles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mobile Injector Unit
Model E 329
For Use With (Equipment) Narrow-Necked Glassware
Includes 4/5 jets 2.5 x 90/110mm, 5/5/5 jets 4.0 x 140/160/180mm, 5/5/5 jets 6.0 x 200/220/240mm, 1 jet for flushing powder dispenser
Quantity 1 Each