Beschreibung
- content: 6 cushions, 3 socks, 20 mats
Spezifikation
Spezifikation
| Material | Heavy-duty Cloth |
| Type | Universal All Purpose Absorbent Storage Cart Refill Kit |
| Weight (English) | 6.61 lb. |
| Weight (Metric) | 3 kg |