Learn More
MK-5046, MedChemExpress
Description
MK-5046 is a novel BRS-3 agonist, binds to BRS-3 with high affinity (mouse Ki = 1.6 nM, human Ki = 25 nM).
Specifications
Specifications
| Chemical Name or Material | MK-5046 |
| Physical Form | Powder |
| CAS | 1022152-70-0 |
| Color | Light Yellow |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H18F6N4O |
| Solubility Information | DMSO : 14.29 mg/mL (32.16 mM; Need ultrasonic) |
| SMILES | O[C@](CC1=NC(CC2(C(F)(F)F)CC2)=CN1)(C(F)(F)F)C3=CC=C(N4C=CC=N4)C=C3 |
| Molecular Weight (g/mol) | 444.37 |
| Show More |