Learn More
MK-3207, MedChemExpress
Product Code. 16483768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483768 10 mg 10mg
16426497 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16483768 Supplier Medchem Express Supplier No. HY10301
MedChemExpress MK-3207 is a potent and orally bioavailable CGRP receptor antagonist (IC50= 0.12 nM; Ki= 0.024 nM); highly selective versus human AM1, AM2, CTR, and AMY3.
MK-3207 is a potent and orally bioavailable CGRP receptor antagonist (IC50= 0.12 nM
Specifications
Chemical Name or Material MK-3207
CAS 957118-49-9
Color White
Purity Grade Notes Research
Molecular Formula C31H29F2N5O3
Quantity 10 mg
Solubility Information DMSO : ≥ 150 mg/mL (269.01 mM)
SMILES O=C1C2(NC[C@@H](C3=CC(F)=CC(F)=C3)N1CC(NC4=CC5=C(C[C@@]6(C(NC7=C6C=CC=N7)=O)C5)C=C4)=O)CCCC2
Molecular Weight (g/mol) 557.59
Formula Weight 557.59
Percent Purity 96.47%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation