missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mizoribine, MedChemExpress
Description
Mizoribine (NSC 289637), an imidazole nucleoside, inhibits HCV RNA replication with IC50 of approximately 100 μM for anti-HCV activity. Immunosuppressant. Mizoribine, an IMPDH inhibitor, inhibits replication of SARS-CoV with IC50s of 3.5 μg/mL and 16 μg/mL for SARS-CoV Frankfurt-1 and SARS-CoV HKU39849, respectively.
Specifications
Specifications
| Chemical Name or Material | Mizoribine |
| CAS | 50924-49-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C9H13N3O6 |
| Quantity | 10 mg |
| Synonym | NSC 289637 HE 69 |
| Solubility Information | DMSO : 125 mg/mL (482.22 mM; Need ultrasonic) |
| SMILES | O[C@@H]1[C@@H](CO)O[C@@H](N2C(O)=C(C(N)=O)N=C2)[C@@H]1O |
| Molecular Weight (g/mol) | 259.22 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?