Learn More
Mixture of (R,S)-N-Nitroso Anabasine-d4 + rac N’-Nitrosonornicotine-d4 + 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone-d4 + (S)-N-Nitroso Anatabine-d4 (0.1mg/mL in Acetonitrile), TRC
Specifications
Specifications
| Chemical Name or Material | (RS)-N-Nitrosoanabasine pyridinyl-d4∣(S)-N-Nitrosoanatabine 2,4,5,6 D4∣N-Nitrosonornicotine 2,4,5,6 D4 (pyridine D4)∣N-Nitrosonornicotine-ketoneD4(pyridylD4) |
| CAS | 1020719-68-9∣1426174-82-4∣66148-19-4∣764661-24-7 |
| Solvent or Matrix | Acetonitrile |
| Molecular Formula | C10 2H4 H9 N3 O∣C10 2H4 H7 N3 O∣C9 2H4 H7 N3 O∣C10 2H4 H9 N3 O2 |
| Quantity | 1x1 mL |
| Synonym | 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine, 2,3,4,6-tetradeuterio-5-[(2S)-1-nitroso-3,6-dihydro-2H-pyridin-2-yl]pyridine, 2,3,4,6-tetradeuterio-5-(1-nitrosopyrrolidin-2-yl)pyridine, N-methyl-N-[4-oxo-4-(2,4,5,6-tetradeuteriopyridin-3-yl)butyl]nitrous amide |
| InChI Formula | InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/i3D,4D,6D,8D∣InChI=1S/C10H11N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h1-4,6,8,10H,5,7H2/t10-/m0/s1/i3D,4D,6D,8D∣InChI=1S/C9H11N3O/c13-11-12-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9H,2,4,6H2/i1D,3D,5D,7D∣InChI=1S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8H,3,5,7H2,1H3/i2D,4D,6D,8D |
| SMILES | [2H]c1nc([2H])c(C2CCCCN2N=O)c([2H])c1[2H]∣[2H]c1nc([2H])c([C@@H]2CC=CCN2N=O)c([2H])c1[2H]∣[2H]c1nc([2H])c(C2CCCN2N=O)c([2H])c1[2H]∣[2H]c1nc([2H])c(C(=O)CCCN(C)N=O)c([2H])c1[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(1-nitrosopiperidin-2-yl)pyridine∣2,3,4,6-tetradeuterio-5-[(2S)-1-nitroso-3,6-dihydro-2H-pyridin-2-yl]pyridine∣2,3,4,6-tetradeuterio-5-(1-nitrosopyrrolidin-2-yl)pyridine∣N-methyl-N-[4-oxo-4-(2,4,5,6-tetradeuteriopyridin-3-yl)butyl]nitrous amide |
| Molecular Weight (g/mol) | 195.25∣193.24∣181.23∣211.25 |
| Show More |