Learn More
Mix 1 of Musk a. Polycy. Musk Comp. 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 33704-61-9, 13171-00-1, 1222-05-5, 83-66-9, 81-14-1, 81-15-2 |
| Quantity | 1 mL |
| Concentration | Cashmeran 10 μg/mL∣Celestolide 10 μg/mL∣HHCB (Galaxolide) 10 μg/mL∣Musk ambrette (technical) 10 μg/mL∣Musk ketone 10 μg/mL∣Musk xylene 10 μg/mL∣MuskNN 10 μg/mL∣Phantolide 10 μg/mL∣Tonalide 10 μg/mL∣Traesolide 10 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Cyclohexane |