Learn More
SP Genevac™ miVac™ Quattro Rotor for Dionex ASE™ 60 mL Vials
Product Code. 17831508
Change view
Click to view available options
Holds:
60 mL Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds
17831508 60 mL Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17831508 Supplier SP Genevac™ Supplier No. QRC28V140030
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Solid Aluminum rotor can hold up to 30 - 60 mL Dionex ASE™ vials.
  • Manufactured from solid aluminum
  • Fits 60 mL Dionex ASE™ vials
  • Can hold up to 30 vials
Specifications
Type Rotor
Holds 60 mL Vials